Skip to Content
Merck
All Photos(1)

Key Documents

SML2072

Sigma-Aldrich

OX03050

≥98% (HPLC)

Synonym(s):

(E)-3-(4-Styrylphenoxy)propan-1-ol, 3-[4-[(1E)-2-Phenylethenyl]phenoxy]-1-propanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€101.00
25 MG
€326.00

€101.00


In StockDetails



Select a Size

Change View
5 MG
€101.00
25 MG
€326.00

About This Item

Empirical Formula (Hill Notation):
C17H18O2
CAS Number:
Molecular Weight:
254.32
UNSPSC Code:
12352200
NACRES:
NA.77

€101.00


In StockDetails


Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

OX03050 is a metabolically stable, potent and selective squalene synthase inhibitor that upregulates the Low-Density Lipoprotein Receptor (LDLR) in mouse and in human liver cell lines. OX03050 used in conjunction with statin acts synergistically to increase LDLR expression
metabolically stable, potent and selective squalene synthase inhibitor that upregulates the LDLR in mouse and in human liver cell lines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alastair G Kerr et al.
The Journal of pharmacology and experimental therapeutics, 361(3), 417-428 (2017-04-01)
Hypercholesterolemia remains one of the leading risk factors for the development of cardiovascular disease. Many large double-blind studies have demonstrated that lowering low-density lipoprotein (LDL) cholesterol using a statin can reduce the risk of having a cardiovascular event by approximately

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service