Skip to Content
Merck
All Photos(1)

Key Documents

SML1809

Sigma-Aldrich

PPTN trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

4-[4-(4-Piperidinyl)phenyl]-7-[4-(trifluoromethyl)phenyl]-2-naphthalenecarboxylic acid trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€195.00
25 MG
€785.00

€195.00


In StockDetails



Select a Size

Change View
5 MG
€195.00
25 MG
€785.00

About This Item

Empirical Formula (Hill Notation):
C29H24F3NO2 · C2HF3O2
CAS Number:
Molecular Weight:
589.52
UNSPSC Code:
12352200
NACRES:
NA.77

€195.00


In StockDetails


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C1=CC2=CC(C3=CC=C(C(F)(F)F)C=C3)=CC=C2C(C4=CC=C(C5CCNCC5)C=C4)=C1)=O.FC(F)(C(O)=O)F

InChI

1S/C29H24F3NO2.C2HF3O2/c30-29(31,32)25-8-5-19(6-9-25)22-7-10-26-23(15-22)16-24(28(34)35)17-27(26)21-3-1-18(2-4-21)20-11-13-33-14-12-20;3-2(4,5)1(6)7/h1-10,15-17,20,33H,11-14H2,(H,34,35);(H,6,7)

InChI key

UAHMBIXBIRLMAG-UHFFFAOYSA-N

Biochem/physiol Actions

PPTN is a P2Y14 receptor antagonist.
PPTN is a very potent (IC50 = 1 nM) P2Y14 receptor antagonist that does not interact with any other P2Y receptor at concentrations up to 10 μM. PPTN blocks UDP-glucose inhibition of forskolin-induced cAMP accumulation, and neutrophil chemotaxis towards UDP-glucose gradients with nM potency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service