Skip to Content
Merck
All Photos(1)

Documents

SML1801

Sigma-Aldrich

CeMMEC13

≥98% (HPLC)

Synonym(s):

N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,2-dihydro-1-methyl-2-oxo-4-quinolinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16N2O4
CAS Number:
Molecular Weight:
336.34
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN(C1=C2C=CC=C1)C(C=C2C(NC3=CC=C4C(OCCO4)=C3)=O)=O

Biochem/physiol Actions

CeMMEC13 is a potent inhibitor of the second bromodomain of the transcription initiation factor TAF1 with an an IC50 value of 2.1 μM and with minimal inhibition of BRD4, BRD9, CREBBP, or EP300. CeMMEC13 was found to synergize with BRD4 inhibitor (S)-JQ1 to arrest the proliferation of leukemia cell line THP1 and lung adenocarcinoma cell line H23.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service