Skip to Content
Merck
All Photos(1)

Key Documents

SML1647

Sigma-Aldrich

Epiblastin A

≥98% (HPLC)

Synonym(s):

2,4,7-Triamino-6-(m-chlorophenyl)-pteridine, 6-(3-Chlorophenyl)-2,4,7-pteridinetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10ClN7
CAS Number:
Molecular Weight:
287.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

NC1=NC(N)=C2C(N=C(N)C(C3=CC=CC(Cl)=C3)=N2)=N1

InChI

1S/C12H10ClN7/c13-6-3-1-2-5(4-6)7-9(14)18-11-8(17-7)10(15)19-12(16)20-11/h1-4H,(H6,14,15,16,18,19,20)

InChI key

ZWNKKZSRANLVEW-UHFFFAOYSA-N

Biochem/physiol Actions

Epiblastin A is triamterene derivative that potently induces reprograming of epiblast stem cells (EpiSC) into embryonic stem cells (ESC). Epiblastin A is a potent reversible inhibitor of casein kinase 1 (CK1) α, δ, and ε that binds to ATP-binding site.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service