Skip to Content
Merck
All Photos(1)

Key Documents

SML1584

Sigma-Aldrich

CMPPE

≥95% (HPLC)

Synonym(s):

2-[1-[2-(4-Chlorophenyl)-5-methylpyrazol[1,5-a]pyrimidin-7-yl]-2-piperidinyl]ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€284.00

€284.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€284.00

About This Item

Empirical Formula (Hill Notation):
C20H23ClN4O
CAS Number:
Molecular Weight:
370.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€284.00


Estimated to ship on15 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1C2=NN(C(N3C(CCO)CCCC3)=CC(C)=N4)C4=C2

InChI

1S/C20H23ClN4O/c1-14-12-20(24-10-3-2-4-17(24)9-11-26)25-19(22-14)13-18(23-25)15-5-7-16(21)8-6-15/h5-8,12-13,17,26H,2-4,9-11H2,1H3

InChI key

TZWSGOPCLUTFBZ-UHFFFAOYSA-N

Biochem/physiol Actions

CMPPE is a GABAB receptor positive allosteric modulator (PAM).
CMPPE is a GABAB receptor positive allosteric modulator (PAM). GABAB receptor positive allosteric modulators have potential for treatment of many neuro-psychiatric disorders as well as drug dependence and seizures. The receptor is a GPCR expressed throughout the CNS; it acts as both an auto-receptor and heteroreceptor to mudulate GABA release and regulate K+ channels and Ca++ channels. CMPPE has the highest hypoactivity/anticonvulsant ratio.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service