Skip to Content
Merck
All Photos(1)

Key Documents

SML1513

Sigma-Aldrich

SR1848

≥98% (HPLC)

Synonym(s):

6-[4-(3-Chlorophenyl)-1-piperazinyl]-3-cyclohexyl-2,4(1H,3H)-pyrimidinedione, CDI 3238389, ML180

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€137.00
25 MG
€553.00

€137.00


Available to ship on27 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€137.00
25 MG
€553.00

About This Item

Empirical Formula (Hill Notation):
C20H25ClN4O2
CAS Number:
Molecular Weight:
388.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€137.00


Available to ship on27 March 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(N(C1CCCCC1)C(N2)=O)C=C2N3CCN(C4=CC(Cl)=CC=C4)CC3

InChI

1S/C20H25ClN4O2/c21-15-5-4-8-17(13-15)23-9-11-24(12-10-23)18-14-19(26)25(20(27)22-18)16-6-2-1-3-7-16/h4-5,8,13-14,16H,1-3,6-7,9-12H2,(H,22,27)

InChI key

CHMQQIFPOIZPIJ-UHFFFAOYSA-N

Application

SR1848 has been used as a liver receptor homolog 1 (LRH-1) inhibitor to study the role of LRH-1 in the regulation of lipopolysaccharide (LPS)-induced proinflammatory cytokines in mice macrophage.[1] It has also been used as an LRH-1 inhibitor to study its effects on activation-induced Fas ligand (FasL) expression in the murine T-cell hybridoma cell line.[2]

Biochem/physiol Actions

SR1848 is a cell penetrant and potent inhibitor of LRH-1.
SR1848 is a cell penetrant and potent inhibitor of orphan nuclear receptor liver receptor homolog 1 (LRH-1; NR5A2) that reduces the expression of CyclinD1 and E1, which results in inhibition of cell proliferation. SR1848 induces rapid translocation of nuclear LRH-1 to the cytoplasm.
The actions of SR1848 might contribute to anti-cancer functionality.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliane Schwaderer et al.
Cell death & disease, 8(4), e2745-e2745 (2017-04-14)
CD95/Fas ligand (FasL) is a cell death-promoting member of the tumor necrosis factor family with important functions in the regulation of T-cell homeostasis and cytotoxicity. In T cells, FasL expression is tightly regulated on a transcriptional level involving a complex
Juliane Schwaderer et al.
Cell death & disease, 11(2), 154-154 (2020-03-01)
Liver receptor homolog-1 (LRH-1, Nr5a2) is an orphan nuclear receptor mainly expressed in tissues of endodermal origin, where its physiological role has been extensively studied. LRH-1 has been implicated in liver cell differentiation and proliferation, as well as glucose, lipid
Cesar A Corzo et al.
Molecular pharmacology, 87(2), 296-304 (2014-12-05)
The orphan nuclear receptor liver receptor homolog 1 (LRH-1; NR5A2) is a potent regulator of cholesterol metabolism and bile acid homeostasis. Recently, LRH-1 has been shown to play an important role in intestinal inflammation and in the progression of estrogen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service