Skip to Content
Merck
All Photos(1)

Key Documents

SML0858

Sigma-Aldrich

SC-1

≥98% (HPLC)

Synonym(s):

N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-(4-cyanophenoxy)phenyl]-urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€98.50

€98.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€98.50

About This Item

Empirical Formula (Hill Notation):
C21H13ClF3N3O2
CAS Number:
Molecular Weight:
431.80
UNSPSC Code:
12352200
NACRES:
NA.77

€98.50


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

SC-1 is a derivative of the multiple tyrosine kinase inhibitor sorafenib with no kinase-inhibition activity. SC-1 blocks STAT3 phosphorylation and activation with similar potency to sorafenib, and induces apoptosis in hepatocellular carcinoma cell lines.
SC-1 is an inhibitor of STAT3 signaling pathway.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye F Tian et al.
Biomaterials, 73, 110-119 (2015-09-26)
Non-Hodgkin lymphomas are a heterogeneous group of lymphoproliferative disorders of B and T cell origin that are treated with chemotherapy drugs with variable success rate that has virtually not changed over decades. Although new classes of chemotherapy-free epigenetic and metabolic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service