Skip to Content
Merck
All Photos(1)

Key Documents

SML0712

Sigma-Aldrich

1-Methyl-1,2,3,4-tetrahydroisoquinoline

≥97% (GC)

Synonym(s):

1,2,3,4-Tetrahydro-1-methylisoquinoline, 1MeTIQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N
CAS Number:
Molecular Weight:
147.22
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (GC)

form

oil

color

, colorless to light yellow to light pink

storage temp.

2-8°C

InChI

1S/C10H13N/c1-8-10-5-3-2-4-9(10)6-7-11-8/h2-5,8,11H,6-7H2,1H3

InChI key

QPILYVQSKNWRDD-UHFFFAOYSA-N

Biochem/physiol Actions

1-Methyl-1,2,3,4-tetrahydroisoquinoline (1MeTIQ) is an endogenous antidepressant and parkinsonism-preventing substance that demonstrates neuroprotectiveactivity.
1-Methyl-1,2,3,4-tetrahydroisoquinoline (1MeTIQ) is an endogenous antidepressant and parkinsonism-preventing substance that demonstrates neuroprotectiveactivity. Following systemic administration in rats, 1-Methyl-1,2,3,4-tetrahydroisoquinoline produces antidepressant-like effect similar to the effect of imipramine. 1MeTIQ is a reversible short-acting moderate inhibitor of MAO A/B.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism and Histamine Synthesis and Metabolism pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Filip et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 58(4), 625-639 (2008-01-16)
The efficacy of 1-methyl-1,2,3,4-tetrahydroisoquinoline (1MeTIQ), a member of endogenous tetrahydroisoquinolines, in cocaine- and food-maintained responding in self-administration procedures under a fixed ratio 5 schedule of reinforcement as well as in cocaine and food seeking behaviors in male Wistar rats was
L Antkiewicz-Michaluk et al.
Journal of neural transmission (Vienna, Austria : 1996), 114(3), 307-317 (2006-08-10)
Drug abuse disorder is induced by a variety of substances and results from their interaction with the brain reward system. It is characterized by a high frequency of relapse, usually associated with to craving. In this study we investigated the
Elhadi Absi et al.
Brain research, 955(1-2), 161-163 (2002-11-07)
1-Methyl-1,2,3,4-tetrahydroisoquinoline (1-MeTIQ), an endogenous monoamine, which prevents the neurotoxic effect of 1-methyl-4-phenylpyridinium ion (MPP(+)) and other endogenous neurotoxins, has been described as being enzymatically formed in the brain by the 1-MeTIQ synthesizing enzyme (1-MeTIQse). In this paper, we report the
K Ishiwata et al.
Journal of neurochemistry, 79(4), 868-876 (2001-11-28)
Parkinsonism-inducing neurotoxicity of 1,2,3,4-tetrahydroisoquinoline (TIQ), as contrasted to 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP), and parkinsonism-preventing effect of 1-methyl-1,2,3,4-tetrahydroisoquinoline (1-MeTIQ) have been investigated in mice by measuring their effects on the in vivo binding of radioligand to pre-synaptic dopamine transporters (DATs) or to dopamine
Yoshie Horiguchi et al.
Chemical & pharmaceutical bulletin, 52(2), 214-220 (2004-02-06)
The sulfoxides 7b and 7d carrying thiophene or benzothiophene as heteroaromatic nucleophiles, when treated with trifluoroacetic anhydride at room temperature (Pummerer reaction), underwent an intramolecular alkylation in an exclusive manner to yield 4,5,6,7-tetrahydro-7-methyl-4-phenylsulfanylthieno[2,3-c]pyridine-6-carbaldehyde (10) and the corresponding benzothiophene derivative (12b)

Articles

Our company offers many products related to histamine synthesis and metabolism for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service