Skip to Content
Merck
All Photos(1)

Key Documents

SML0204

Sigma-Aldrich

MS-1020

≥98% (HPLC)

Synonym(s):

1-Hydroxy-N-(2-(5-hydroxy-1H-indol-3-yl)ethyl)-2-naphtamide, 1-Hydroxy-N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]-2-naphthalenecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€237.00

€237.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€237.00

About This Item

Empirical Formula (Hill Notation):
C21H18N2O3
CAS Number:
Molecular Weight:
346.38
UNSPSC Code:
12352202
NACRES:
NA.77

€237.00


Estimated to ship on31 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥10 mg/mL at 60 °C

storage temp.

2-8°C

Application

MS-1020 may be used in JAK-3-mediated cell signaling studies.

Biochem/physiol Actions

MS-1020 is a selective JAK3 inhibitor
MS-1020 is an ATP-competitive, selective inhibitor of JAK3, thereby blocking downstream STAT signaling. MS-1020 binds directly with JAK3, blocking its catalytic activity. It suppressed IL-2-induced JAK3/STAT5 signaling but not prolactin-induced JAK2/STAT5 signaling. MS-1020 has also been shown to decreases cell survival by inducing apoptosis via downregulating anti-apoptotic gene expression.

Features and Benefits

This compound is featured on the JAKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service