Skip to Content
Merck
All Photos(1)

Key Documents

SCP0254

Sigma-Aldrich

TNF Coverting Enzyme Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€255.00

€255.00


Check Cart for Availability


Select a Size

Change View
1 MG
€255.00

About This Item

Empirical Formula (Hill Notation):
C28H32N4O7
Molecular Weight:
536.58
UNSPSC Code:
12352202
NACRES:
NA.32

€255.00


Check Cart for Availability

Product Name

TNF Coverting Enzyme Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥85%

storage condition

protect from light

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SCP0257SCP0105SCP0066
form

lyophilized

form

lyophilized

form

lyophilized

form

lyophilized

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage condition

protect from light

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

composition

Peptide Content, ≥85%

composition

Peptide Content, ≥65%

composition

Peptide Content, ≥75%

composition

Peptide Content, ≥80%

Amino Acid Sequence

MCA-Pro-Leu-Ala-Gln-Ala-Val-DNP-Dap-Arg-Ser-Ser-Ser-Arg-NH2

Application

TNF Coverting Enzyme (TACE) is responsible for shedding of various membrane proteins including proinflammatory cytokine TNF. MCA-PLAQAV-DNP-Dap-RSSSR-NH2 is used as a TACE substrate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service