Skip to Content
Merck
All Photos(1)

Key Documents

SCP0046

Sigma-Aldrich

Amyloid β 28-35 human

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€109.00
5 MG
€366.00

€109.00


Estimated to ship on28 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€109.00
5 MG
€366.00

About This Item

Empirical Formula (Hill Notation):
C36H67N9O9S1
Molecular Weight:
802.04
UNSPSC Code:
12352200
NACRES:
NA.32

€109.00


Estimated to ship on28 April 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met

Application

Amyloid β (Aβ) refers to peptides derived from Amyloid precursor protein that vary in length from 36-43 amino acids. Aβ(s) peptides, their peptide fragments and mutated fragments are used to study a wide range of metabolic and regulatory functions including activation of kinases, regulation of cholesterol transport, function as a transcription factor, and regulators of inflammation. Aβ(s) peptides and their peptide fragments are also used to study oxidative stress, metal binding and mechanisms of protein cross-linking in the context of diseases such as Alzheimer′s disease and neurodegeneration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sureshbabu Nagarajan et al.
The FEBS journal, 275(10), 2415-2427 (2008-04-22)
The interaction of the beta-amyloid peptide (Abeta) with neuronal membranes could play a key role in the pathogenesis of Alzheimer's disease. Recent studies have focused on the interactions of Abeta oligomers to explain the neuronal toxicity accompanying Alzheimer's disease. In
Sureshbabu Nagarajan et al.
Journal of structural biology, 170(3), 439-450 (2010-03-02)
Folding and self-assembly of amyloid beta (Abeta) peptide are linked to Alzheimer's disease. To understand the initial stage of amyloid-beta peptide aggregation, conformational characteristics of monomers of wild-type (WT) Abeta(28-35) and its mutant peptides A30G and A30I were investigated using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service