Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702599

Sigma-Aldrich

Anti-NEK2 Antibody, clone 5O0A1, Rabbit Monoclonal

Synonym(s):

HsPK21, NEK2A, NLK1, PPP1R111, RP67

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€378.00

€378.00


Estimated to ship on11 April 2025



Select a Size

Change View
100 μL
€378.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€378.00


Estimated to ship on11 April 2025


biological source

rabbit

Quality Level

material

colorless

clone

5O0A1, monoclonal

form

liquid

mol wt

50 kDa

species reactivity

human, rat, rat

concentration

0.7 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MPSRAEDYEVLYTIGTGSYGRCQKIRRKSDGKILVWKELDYGSMTEAEKQMLVSEVNLLRELKHPNIVRYYDRIIDRTNTTLYIVMEYCEGGDLASVITK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEK2(4751)

General description

This gene encodes a serine/threonine-protein kinase that is involved in mitotic regulation. This protein is localized to the centrosome, and undetectable during G1 phase, but accumulates progressively throughout the S phase, reaching maximal levels in late G2 phase. Alternatively spliced transcript variants encoding different isoforms with distinct C-termini have been noted for this gene. [provided by RefSeq, Feb 2011]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NEK2 (P51955).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service