Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702264

Sigma-Aldrich

Anti-TIMP2 Antibody, clone 8R5Q2, Rabbit Monoclonal

Synonym(s):

TIMP2,CSC-21K,DDC8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

material

colorless

clone

8R5Q2, monoclonal

form

liquid

mol wt

22 kDa

species reactivity

African green monkey, rat, mouse, human

concentration

1.00 mg/mL

technique(s)

western blot: 1:500 - 1:1000

color

colorless

isotype

IgG

immunogen sequence

VSLDVGGKKEYLIAGKAEGDGKMHITLCDFIVPWDTLSTTQKKSLNHRYQMGCECKITRCPMIPCYISSPDECLWMDWVTEKNINGHQAKFFACIKRSDGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMP2(7077)

General description

This gene is a member of the TIMP gene family. The proteins encoded by this gene family are natural inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. In addition to an inhibitory role against metalloproteinases, the encoded protein has a unique role among TIMP family members in its ability to directly suppress the proliferation of endothelial cells. As a result, the encoded protein may be critical to the maintenance of tissue homeostasis by suppressing the proliferation of quiescent tissues in response to angiogenic factors, and by inhibiting protease activity in tissues undergoing remodelling of the extracellular matrix.

Immunogen

PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant protein of human TIMP2.

Application

WB

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service