Skip to Content
Merck
All Photos(2)

Key Documents

SAB5700436

Sigma-Aldrich

Anti-Phospho-eIF2α-S51 antibody produced in rabbit

Synonym(s):

EIF-2, EIF-2A, EIF-2alpha, EIF2, EIF2A, EIF2S1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€315.00

€315.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1419


Select a Size

Change View
100 μL
€315.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€315.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1419

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36

species reactivity

human

concentration

0.77 mg/ml

technique(s)

immunoprecipitation (IP): 1:50-1:100
western blot: 1:1000-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer51)

Gene Information

human ... EIF2S1(1965)

Immunogen

A synthetic phosphorylated peptide around S51 of human eIF2α (NP_004085.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service