Skip to Content
Merck
All Photos(1)

Key Documents

SAB5700388

Sigma-Aldrich

Anti-Phospho-HER2/ERBB2-Y1221/1222 antibody produced in rabbit

Synonym(s):

CD340, ERBB2, ErbB2, HER-2, HER-2/neu, HER2, HER2/ErbB2, MLN 19, MLN19, NEU, NGL, TKR1, erb-b2 receptor tyrosine kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€371.00

€371.00


Estimated to ship on15 May 2025



Select a Size

Change View
100 μL
€371.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€371.00


Estimated to ship on15 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

134
136
137
62
70
97

species reactivity

human

concentration

5.8 mg/ml

technique(s)

western blot: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1221/pTyr1222)

Gene Information

human ... ERBB2(2064)

General description

This gene encodes a member of the epidermal growth factor (EGF) receptor family of receptor tyrosine kinases. This protein has no ligand binding domain of its own and therefore cannot bind growth factors. However, it does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signalling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Allelic variations at amino acid positions 654 and 655 of isoform a (positions 624 and 625 of isoform b) have been reported, with the most common allele, Ile654/Ile655, shown here. Amplification and/or overexpression of this gene has been reported in numerous cancers, including breast and ovarian tumors. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.

Immunogen

A synthetic phosphorylated peptide around Y1221 & Y1222 of human HER2/ERBB2 (NP_004439.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service