Skip to Content
Merck
All Photos(3)

Key Documents

SAB5700310

Sigma-Aldrich

Anti-Phospho-MAP2K1-S298 antibody produced in rabbit

Synonym(s):

CFC3, MAP2K1, MAPKK1, MEK1, MKK1, PRKMK1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€371.00

€371.00


Estimated to ship on08 May 2025



Select a Size

Change View
100 μL
€371.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€371.00


Estimated to ship on08 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40
43

species reactivity

rat, human, mouse

concentration

0.94 mg/ml

technique(s)

immunoprecipitation (IP): 1:50-1:100
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer298)

Gene Information

human ... MAP2K1(5604)

General description

The protein encoded by this gene is a member of the dual specificity protein kinase family, which acts as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein kinase lies upstream of MAP kinases and stimulates the enzymatic activity of MAP kinases upon wide variety of extra- and intracellular signals. As an essential component of MAP kinase signal transduction pathway, this kinase is involved in many cellular processes such as proliferation, differentiation, transcription regulation and development.

Immunogen

A synthetic phosphorylated peptide around S298 of human MAP2K1 (NP_002746.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service