Skip to Content
Merck
All Photos(2)

Key Documents

SAB4301579

Sigma-Aldrich

Anti-KIR6.2 (PHOSPHO-THR224)

affinity isolated antibody

Synonym(s):

ATP-sensitive inward rectifier potassium channel 11, IKATP, IRK11, Inward rectifier K channel Kir6.2, KCNJ11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€326.00

€326.00


Estimated to ship on15 May 2025



Select a Size

Change View
100 μL
€326.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€326.00


Estimated to ship on15 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human, mouse

technique(s)

immunofluorescence: 1:100-1:500
western blot: 1:500-1:3000

accession no.

AC124798

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr224)

Gene Information

human ... KCNJ11(3767)

Immunogen

Peptide sequence around phosphorylation site of threonine 224 (K-T-TP-S-P) derived from Human Kir6.2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service