Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300232

Sigma-Aldrich

Anti-phospho-GRIA2 (pSer880) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GLUR2 antibody produced in rabbit, Anti-GLURB antibody produced in rabbit, Anti-GluR-K2 antibody produced in rabbit, Anti-HBGR2 antibody produced in rabbit, Anti-glutamate receptor, ionotropic, AMPA 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~100 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(I-E-SP-V-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer880)

Gene Information

human ... GRIA2(2891)

Immunogen

Peptide sequence around phosphorylation site of serine 880 (I-E-S(p)-V-K), according to the protein GRIA2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Ionotropic glutamate receptor. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter L-glutamate induces a conformation change, leading to the opening of the cation channel, and thereby converts the chemical signal to an electrical impulse. The receptor then desensitizes rapidly and enters a transient inactive state, characterized by the presence of bound agonist.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahima Sharma et al.
Cerebral cortex (New York, N.Y. : 1991), 29(7), 2932-2946 (2018-07-10)
Metabotropic glutamate receptor-dependent long-term depression (mGluR-LTD) is conventionally considered to be solely dependent on local protein synthesis. Given the impact of epigenetics on memory, the intriguing question is whether epigenetic regulation influences mGluR-LTD as well. G9a/GLP histone lysine methyltransferase complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service