Skip to Content
Merck
All Photos(1)

Documents

SAB4200855

Sigma-Aldrich

Anti-Cytokeratin Peptide 18 antibody, Mouse monoclonal

clone CY-90, purified from hybridoma cell culture

Synonym(s):

Anti-CK18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

antibody form

purified from hybridoma cell culture

Quality Level

antibody product type

primary antibodies

clone

CY-90, monoclonal

form

liquid

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: 10-20 μg/mL using heat-retrieved formalin-fixed, paraffin-embedded human placenta sections

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Intermediate filaments (IFs) are one of three major cytoskeletal filament assemblies in higher eukaryotes.6 Keratins (formerly known as “cytokeratins”) comprise of types I and II IF genes (given a total of 54 conserved genes) that are highly expressed primarily in epithelial cells hair and skin appendages. Cytokeratins are the largest subgroup of IF in mammals and are the predominant subtype of IFs in all epithelia.6,7 Cytokeratins form a 10-nm filaments comprised of type I and II cytokeratine heteropolymer.7

Specificity

Monoclonal Anti-Cytokeratin Peptide 18 antibody specifically recognizes cytokeratin peptide 18 in human cultured cell lines as well as human tissue extracts. The antibody also shows reactivity with samples from rat1 and monkey2 origin. The antibody reacts specifically with a wide variety of simple epithelia (e.g. intestine, respiratory and urinary systems, liver and glandular epithelia). It does not react with stratified squamous epithelia (e.g. esophagus or epidermis) or with non-epithelial cells.

Application

The antibody may be used in various immunochemical techniques including Immunoblotting (~45 kDa)3, Immunofluorescence1-2, Immunoprecipitation4 and Immunohistology5.

Biochem/physiol Actions

Cytokeratins provide the cells with structural support, resistance to chemical stresses and maintenance of a polarized cytoarchitecture.6 Each epithelial tissue has a specific expression pattern of cytokeratins, which makes them common and excellent pathology markers for tumor identification, classification and epithelial cell differentiation.6,7
Cytokeratin 18 is a member of the type I subfamily. Cytokeratin pair 8/18 forms the IF scaffold predominantly in simple epithelia such as the liver and pancreas. They have major role in the mechanical support of the cell, protein biosynthesis, protection from apoptosis, regulation of cell cycle progression, motility and organelle transport.3
Cytokeratins over and misexpression are linked to various diseases and more than 60 different disorders (termed as keratinopathies) have been linked to inherited cytokeratin changes. Cytokeratins (such as cytokeratin 18) serve as serum markers for malignant and non-neoplastic disorders and mutation of human cytokeratin 18 has also been associated with liver diseases.7

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative. Antibody Concentration: ~ 1.0 mg/mL

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service