Skip to Content
Merck
All Photos(3)

Key Documents

SAB3501242

Sigma-Aldrich

Anti-SCARB1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
WB
clone:
polyclonal
species reactivity:
rat, human, mouse
citations:
technique(s):
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCARB1(949)

General description

SCARB1 Antibody: Scavenger receptor class B member 1 (SCARB1), also known as SR-BI, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARB1 is a multi-ligand cell-surface receptor that mediates the selective uptake of lipid from HDL cholesterol into cells and is expressed in steroidogenic tissues in adult animals. Other ligands of SCARB1 include native, acetylated, or oxidized LDL and anionic phospholipids. SCARB1-deficient mice have elevated HDL levels and increased susceptibility to atherosclerosis on fat feeding, indicating its importance in the regulation of cholesterol homeostasis. Along with CLDN1, LDL-R, and the tetraspanin superfamily member CD81, SCARB1 has been reported to be an entry factor for the Hepatitis C virus.

Immunogen

SCARB1 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human SCARB1.The immunogen is located within amino acids 70 - 120 of SCARB1.

Application

SCARB1 antibody can be used for detection of SCARB1 by Western blot at 1 - 2 μg/mL. Antibody can also be used for immunohistochemistry starting at 2.5 μg/mL. For immunofluorescence start at 20 μg/mL.Antibody validated: Western Blot in human samples; Immunohistochemistry in human samples and Immunofluorescence in human samples. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service