Skip to Content
Merck
All Photos(2)

Key Documents

SAB2102009

Sigma-Aldrich

Anti-RIPK3 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-RIP3, Anti-Receptor-interacting serine-threonine kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

57 kDa

species reactivity

horse, rat, bovine, human, mouse

technique(s)

western blot: suitable

immunogen sequence

FRAQHRKWGYDVAVKIVNSKAISREVKAMASLDNEFVLRLEGVIEKVNWD

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIPK3(11035)

Immunogen

Peptide region of the protein sequence according to NP_006862.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuo Hojo et al.
Scientific reports, 6, 38211-38211 (2016-12-03)
Programmed necrosis (necroptosis) is an alternative form of programmed cell death that is regulated by receptor-interacting protein kinase (RIPK) 1 and 3-dependent, but is a caspase (CASP)-independent pathway. In the present study, to determine if necroptosis participates in bovine structural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service