Skip to Content
Merck
All Photos(4)

Key Documents

SAB1411064

Sigma-Aldrich

Anti-NLRP2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CLR19.9, FLJ20510, NALP2, NBS1, PAN1, PYPAF2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€417.00

€417.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μG
€417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€417.00


Estimated to ship on30 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 120.5 kDa

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NLRP2(55655)

General description

NALP proteins, such as NALP2, are characterized by an N-terminal pyrin (MIM 608107) domain (PYD) and are involved in the activation of caspase-1 (CASP1; MIM 147678) by Toll-like receptors (see TLR4; MIM 603030). They may also be involved in protein complexes that activate proinflammatory caspases (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM

Immunogen

NLRP2 (NP_060322.1, 1 a.a. ~ 1062 a.a) full-length human protein.

Sequence
MVSSAQMGFNLQALLEQLSQDELSKFKYLITTFSLAHELQKIPHKEVDKADGKQLVEILTTHCDSYWVEMASLQVFEKMHRMDLSERAKDEVREAALKSFNKRKPLSLGITRKERPPLDVDEMLERFKTEAQAFTETKGNVICLGKEVFKGKKPDKDNRCRYILKTKFREMWKSWPGDSKEVQVMAERYKMLIPFSNPRVLPGPFSYTVVLYGPAGLGKTTLAQKLMLDWAEDNLIHKFKYAFYLSCRELSRLGPCSFAELVFRDWPELQDDIPHILAQARKILFVIDGFDELGAAPGALIEDICGDWEKKKPVPVLLGSLLNRVMLPKAALLVTTRPRALRDLRILAEEPIYIRVEGFLEEDRRAYFLRHFGDEDQAMRAFELMRSNAALFQLGSAPAVCWIVCTTLKLQMEKGEDPVPTCLTRTGLFLRFLCSRFPQGAQLRGALRTLSLLAAQGLWAQTSVLHREDLERLGVQESDLRLFLDGDILRQDRVSKGCYSFIHLSFQQFLTALFYTLEKEEEEDRDGHTWDIGDVQKLLSGVERLRNPDLIQAGYYSFGLANEKRAKELEATFGCRMSPDIKQELLRCDISCKGGHSTVTDLQELLGCLYESQEEELVKEVMAQFKEISLHLNAVDVVPSSFCVKHCRNLQKMSLQVIKENLPENVTASESDAEVERSQDDQHMLPFWTDLCSIFGSNKDLMGLAINDSFLSASLVRILCEQIASDTCHLQRVVFKNISPADAHRNLCLALRGHKTVTYLTLQGNDQDDMFPALCEVLRHPECNLRYLGLVSCSATTQQWADLSLALEVNQSLTCVNLSDNELLDEGAKLLYTTLRHPKCFLQRLSLENCHLTEANCKDLAAVLVVSRELTHLCLAKNPIGNTGVKFLCEGLRYPECKLQTLVLWNCDITSDGCCDLTKLLQEKSSLLCLDLGLNHIGVKGMKFLCEALRKPLCNLRCLWLWGCSIPPFSCEDLCSALSCNQSLVTLDLGQNPLGSSGVKMLFETLTCSSGTLRTLRLKIDDFNDELNKLLEEIEEKNPQLIIDTEKHHPWAERPSSHDFMI

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sangeetha Mahadevan et al.
Scientific reports, 7, 44667-44667 (2017-03-21)
Mammalian parental genomes contribute differently to early embryonic development. Before activation of the zygotic genome, the maternal genome provides all transcripts and proteins required for the transition from a highly specialized oocyte to a pluripotent embryo. Depletion of these maternally-encoded

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service