Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410395

Sigma-Aldrich

Anti-CARD8 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CARDINAL, DACAR, DKFZp779L0366, Dakar, FLJ18119, FLJ18121, KIAA0955, MGC57162, NDPP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 44 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CARD8(22900)

General description

The protein encoded by this gene belongs to the caspase recruitment domain (CARD)-containing family of proteins, which are involved in pathways leading to activation of caspases or nuclear factor kappa-B (NFKB). This protein may be a component of the inflammasome, a protein complex that plays a role in the activation of proinflammatory caspases. It is thought that this protein acts as an adaptor molecule that negatively regulates NFKB activation, CASP1-dependent IL1B secretion, and apoptosis. Polymorphisms in this gene may be associated with a susceptibility to rheumatoid arthritis. Alternatively spliced transcript variants have been described for this gene, but their biological validity has not been determined. (provided by RefSeq)

Immunogen

CARD8 (AAH56891.1, 1 a.a. ~ 392 a.a) full-length human protein.

Sequence
MEKKECPEKSSSSEEELPRRDSGSSRNIDASKLIRLQGSRKLLVDNSIRELQYTKTGIFFQAEACVTNDTVYRELPCVSETLCDISHFFQEDDETEAEPLLFRAVPECQLSGGDIPSVSEEQESSEGQDSGDICSEENQIVSSYASKVCFEIEEDYKNRQFLGPEGNVDVELIDKSTNRYSVWFPTAGWYLWSATGLGFLVRDEVTVTIAFGSWSQHLALDLQHHEQWLVGGPLFDVTAEPEEAVAEIHLPHSISLQAGEVDVSWFLVAHFKNEGMVLEHPARVEPFYAVLESPSFSLMGILLRIASGTRLSIPITSNTLIYYHPHPEDIKFHLYLVPSDALLTKAIDDEEDRFHGVRLQTSPPMEPLNFGSSYIVSNSANLKVMPKWISSL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service