Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406297

Sigma-Aldrich

Anti-PRKAR2B antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

PRKAR2, RII-BETA

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€368.00

€368.00


Check Cart for Availability


Select a Size

Change View
50 μG
€368.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€368.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~46.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKAR2B(5577)

General description

cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase, which transduces the signal through phosphorylation of different target proteins. The inactive kinase holoenzyme is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. This subunit has been shown to interact with and suppress the transcriptional activity of the cAMP responsive element binding protein 1 (CREB1) in activated T cells. Knockout studies in mice suggest that this subunit may play an important role in regulating energy balance and adiposity. The studies also suggest that this subunit may mediate the gene induction and cataleptic behavior induced by haloperidol. (provided by RefSeq)

Immunogen

PRKAR2B (AAH75800.1, 1 a.a. ~ 418 a.a) full-length human protein.

Sequence
MSIEIPAGLTELLQGFTVEVLRHQPADLLEFALQHFTRLQQENERKGTARFCHEGRTWGDLGAAAGGGTPSKGVNFAEEPMQSDSEDGEEEEAAPADAGAFNAPVINRFTRRASVCAEAYNPDEEEDDAESRIIHPKTDDQRNRLQEACKDILLFKNLDPEQMSQVLDAMFEKLVKDGEHVIDQGDDGDNFYVIDRGTFDIYVKCDGVGRCVGNYDNRGSFGELALMYNTPRAATITATSPGALWGLDRVTFRRIIVKNNAKKRKMYESFIESLPFLKSLEFSERLKVVDVIGTKVYNDGEQIIAQGDSADSFFIVESGEVKITMKRKGKSEVEENGAVEIARCSRGQYFGELALVTNKPRAASAHAIGTVKCLAMDVQAFERLLGPCMEIMKRNIATYEEQLVALFGTNMDIVEPTA

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service