Skip to Content
Merck
All Photos(4)

Documents

SAB1405990

Sigma-Aldrich

Anti-SP110 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ22835, IFI41, IFI75, IPR1, VODI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~61.8 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SP110(3431)

General description

The nuclear body is a multiprotein complex that may have a role in the regulation of gene transcription. This gene is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. In addition, it has been suggested that the protein may play a role in ribosome biogenesis and in the induction of myeloid cell differentiation. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. (provided by RefSeq)

Immunogen

SP110 (AAH19059.1, 1 a.a. ~ 547 a.a) full-length human protein.

Sequence
MFTMTRAMEEALFQHFMHQKLGIAYAIHKPFPFFEGLLDNSIITKRMYMESLEACRNLIPVSRVVHNILTQLERTFNLSLLVTLFSQINLREYPNLVTIYRSFKRVGASYERQSRDTPILLEAPTGLAEGSSLHTPLALPPPQPPQPSCSPCAPRVSEPGTSSQQSDEILSESPSPSDPVLPLPALIQEGRSTSVTNDKLTSKMNAEEDSEEMPSLLTSTVQVASDNLIPQIRDKEDPQEMPHSPLGSMPEIRDNSPEPNDPEEPQEVSSTPSDKKGKKRKRCIWSTPKRRHKKKSLPRALSRHGIQKKLKRVDQVPQKKDDSTCNSTVETRAQKARTECARKSRSEEIIDGTSEMNEGKRSQKTPSTPRRVTQGAASPGHGIQEKLQVVDKVTQRKDDSTWNSEVMMRVQKARTKCARKSRSKEKKKEKDICSSSKRRFQKNIHRRGKPKSDTVDFHCSKLPVTCGEAKGILYKKKMKHGSSVKCIRNEDGTWLTPNEFEVEGKGRNAKNWKRNIRCEGMTLGELLKSGLLLCPPRINLKRELNSK

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service