Skip to Content
Merck
All Photos(2)

Key Documents

SAB1405143

Sigma-Aldrich

Monoclonal Anti-ARMC4, (C-terminal) antibody produced in mouse

clone 5F1, ascites fluid

Synonym(s):

DKFZp434P1735, FLJ10376, FLJ10817, FLJ32798, RP11-691I13.1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€490.00

€490.00


Estimated to ship on28 April 2025



Select a Size

Change View
200 μL
€490.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€490.00


Estimated to ship on28 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5F1, monoclonal

mol wt

antigen ~37.11 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARMC4(55130)

Related Categories

Immunogen

ARMC4 (NP_060546, 945 a.a. ~ 1044 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
AISRCCMWGRNRVAFGEHKAVAPLVRYLKSNDTNVHRATAQALYQLSEDADNCITMHENGAVKLLLDMVGSPDQDLQEAAAGCISNIRRLALATEKARYT

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service