Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300444

Sigma-Aldrich

Anti-TRRAP (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-TRRAP protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRRAP(8295)

General description

TRRAP, a member of the TRA1 subfamily of the PI3/PI4 kinase family, is an adapter protein found in various multiprotein chromatin complexes with histone acetyltransferase activity (HAT), which gives a specific tag for epigenetic transcription activation. It plays a central role in MYC (c-Myc) transcription activation, and also participates in cell transformation by MYC. TRRAP is required for p53/TP53-, E2F1- and E2F4-mediated transcription activation, and is also involved in transcription activation mediated by the adenovirus E1A, a viral oncoprotein that deregulates transcription of key genes. It likely performs by linking transcription factors such as E1A, MYC or E2F1 to HAT complexes such as STAGA thereby allowing transcription activation. Although probably not required in the steps following histone acetylation in processes of transcription activation, it may be required for the mitotic checkpoint and normal cell cycle progression. It is a component of various HAT complexes, including the PCAF, TFTC, TIP60 STAGA and BAF53 complexes. The PI3K/PI4K domain of this nuclear protein is required for the recruitment of HAT complexes, and the MYC-dependent transactivation. Although it is strongly related to the PI3/PI4-kinase family, it lacks the typical motifs that constitute the catalytic site of PI3/PI4-kinase proteins, and lacks such activity.

Immunogen

TRRAP (Q9Y6H4, 3804-3840)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human TRRAP.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dilraj Lama et al.
Nature communications, 15(1), 1865-1865 (2024-03-01)
The c-MYC oncogene is activated in over 70% of all human cancers. The intrinsic disorder of the c-MYC transcription factor facilitates molecular interactions that regulate numerous biological pathways, but severely limits efforts to target its function for cancer therapy. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service