Skip to Content
Merck
All Photos(1)

Key Documents

S7070

Sigma-Aldrich

Anti-Semaphorin 6A antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Sema 6A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€679.00

€679.00


Estimated to ship on01 June 2025



Select a Size

Change View
100 μG
€679.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€679.00


Estimated to ship on01 June 2025


biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

direct ELISA: 0.5-1 μg/mL using rmWnt-1
immunohistochemistry: 2-5 μg/mL
western blot: 0.1-0.2 μg/mL using rmWnt-1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Sema6a(20358)

Specificity

This antibody shows ~25% cross-reactivity with rhSema 6A and less than 1% cross-reactivity with rhSema 3A, rhSema 3B, rmSema 3C, rmSema 3F, and rmSema 7A.

Immunogen

Sf21-derived, recombinant mouse semaphorin 6A (rmSema 6A).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service