Skip to Content
Merck
All Photos(1)

Key Documents

RAB0415

Sigma-Aldrich

Resistin EIA Kit

for serum, plasma, culture supernatant and cell lysates

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€500.00

€500.00


Estimated to ship on12 May 2025



Select a Size

Change View
1 KIT
€500.00

About This Item

UNSPSC Code:
41123300
NACRES:
NA.32

€500.00


Estimated to ship on12 May 2025


species reactivity

mouse, rat, human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type cell lysate
sample type culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 466 pg/mL
standard curve range: 0.1-1000 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RETN(56729)

General description

The Resistin Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting Resistin peptide based on the principle of Competitive Enzyme Immunoassay. EIA-RES-1 detects Resistin (90aa).

Immunogen

Resistin, synthetic peptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service