Skip to Content
Merck
All Photos(1)

Key Documents

PZ0377

Sigma-Aldrich

PF-05236216 hydrochloride

≥98% (HPLC)

Synonym(s):

4-[3-(4-Fluorophenyl)-1-methyl-1H-pyrazol-4-yl]-6,7-dihydro-6-methyl-5H-pyrrolo[3,4-b]pyridin-5-one hydrochloride, PF-5236216 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€87.30
25 MG
€284.00

€87.30


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€87.30
25 MG
€284.00

About This Item

Empirical Formula (Hill Notation):
C18H15FN4O · HCl
CAS Number:
Molecular Weight:
358.80
UNSPSC Code:
51111800
NACRES:
NA.77

€87.30


Available to ship on31 March 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1C=C(C2=CC=NC(CN3C)=C2C3=O)C(C4=CC=C(F)C=C4)=N1.Cl

InChI

1S/C18H15FN4O.ClH/c1-22-10-15-16(18(22)24)13(7-8-20-15)14-9-23(2)21-17(14)11-3-5-12(19)6-4-11;/h3-9H,10H2,1-2H3;1H

InChI key

CLDJMFXSCFWXDI-UHFFFAOYSA-N

Biochem/physiol Actions

PF-05236216 is a brain penetrant, potent and selective inhibitor of casein kinase 1 delta/epsilon (CK1δ/ε) that modulates circadian rhythms in mice.
brain penetrant, potent and selective inhibitor of casein kinase 1 delta/epsilon (CK1δ/ε)

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scot Mente et al.
Journal of medicinal chemistry, 56(17), 6819-6828 (2013-08-08)
Casein kinase 1δ (CK1δ) and 1ε (CK1ε) are believed to be necessary enzymes for the regulation of circadian rhythms in all mammals. On the basis of our previously published work demonstrating a CK1ε-preferring compound to be an ineffective circadian clock
Travis T Wager et al.
ACS chemical neuroscience, 8(9), 1995-2004 (2017-06-14)
To enable the clinical development of our CNS casein kinase 1 delta/epsilon (CK1δ/ε) inhibitor project, we investigated the possibility of developing a CNS positron emission tomography (PET) radioligand. For this effort, we focused our design and synthesis efforts on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service