Skip to Content
Merck
All Photos(1)

Key Documents

PZ0346

Sigma-Aldrich

PF-06726304 acetate

≥98% (HPLC)

Synonym(s):

5,8-Dichloro-2-[(1,2-dihydro-4,6-dimethyl-2-oxo-3-pyridinyl)methyl]-7-(3,5-dimethyl-4-isoxazolyl)-3,4-dihydro-1(2H)-isoquinolinone acetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€108.00

€108.00


Check Cart for Availability


Select a Size

Change View
5 MG
€108.00

About This Item

Empirical Formula (Hill Notation):
C22H21Cl2N3O3 · C2H4O2
CAS Number:
Molecular Weight:
506.38
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€108.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC(O)=O.CC(ON=C1C)=C1C2=CC(Cl)=C(CCN(CC3=C(C)C=C(C)NC3=O)C4=O)C4=C2Cl

Biochem/physiol Actions

PF-06726304 is a selective inhibitor of Histone-lysine N-methyltransferase EZH2 (enhancer of Zeste homolog 2), which catalyzes trimethylation of histone H3 lysine 27 (H3K27) and is amplified and/or overexpressed in several human cancers including breast, prostate and lymphoma. PF-06726304 is a novel structural class of EZH2 inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service