Skip to Content
Merck
All Photos(1)

Key Documents

PZ0223

Sigma-Aldrich

PF-04991532

≥98% (HPLC)

Synonym(s):

6-[[(2S)-3-Cyclopentyl-1-oxo-2-[4-(trifluoromethyl)-1H-imidazol-1-yl]propyl]amino]-3-pyridinecarboxylic acid, PF 04991532

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€96.90
25 MG
€288.00

€96.90


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€96.90
25 MG
€288.00

About This Item

Empirical Formula (Hill Notation):
C18H19F3N4O3
CAS Number:
Molecular Weight:
396.36
UNSPSC Code:
51111800
NACRES:
NA.77

€96.90


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

FC(F)(F)c1nc[n](c1)[C@@H](CC3CCCC3)C(=O)Nc2ncc(cc2)C(=O)O

InChI

1S/C18H19F3N4O3/c19-18(20,21)14-9-25(10-23-14)13(7-11-3-1-2-4-11)16(26)24-15-6-5-12(8-22-15)17(27)28/h5-6,8-11,13H,1-4,7H2,(H,27,28)(H,22,24,26)/t13-/m0/s1

InChI key

GKMLFBRLRVQVJO-ZDUSSCGKSA-N

Biochem/physiol Actions

PF-04991532 is a potent and hepatoselective glucokinase activator that reduces mean daily glucose (MDG), fasting plasma glucose (FPG) and glucose excursion in humans.
PF-04991532 is a potent and hepatoselective glucokinase activator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service