Skip to Content
Merck
All Photos(4)

Key Documents

PLA0308

Sigma-Aldrich

Goat anti-RelA Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_068810.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... RelA(5970)

Immunogen

The epitope recognized by PLA0308 maps to a region between residue 501 to 551 of human Reticuloendotheliosis Viral Oncogene Homolog A using the numbering given in entry NP_068810.2 (GeneID 5970).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

RelA, is one of five monomeric subunits that are part of the Rel family of proteins that dimerize to form the NF-kappa B (nuclear factor that binds the kappa immunoglobulin light chain gene enhancer) transcription factor. Members of the Rel family of proteins include NF-kappa B1/p50, NF-kappa B2/p52, RelA/p65, c-Rel, and RelB. Each Rel member bears a characteristic 300 amino acid N-terminal Rel homology domain (RHD) that functions to mediate DNA binding, dimerization, nuclear localization, and inhibitor association.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service