Skip to Content
Merck
All Photos(2)

Key Documents

PLA0039

Sigma-Aldrich

Goat anti-MCM4 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CDC21, CDC21 homolog, CDC54, MCM4 minichromosome maintenance deficient 4 (S. cerevisiae), MGC33310, NKCD, NKGCD, P1-Cdc21, hCdc21, homolog of S. pombe cell devision cycle 21, minichromosome maintenance deficient 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€470.00

€470.00


Estimated to ship on30 April 2025



Select a Size

Change View
100 μL
€470.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€470.00


Estimated to ship on30 April 2025


biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:500-1:2,000
western blot: 1:2,000-1:10,000

accession no.

NP_005905.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

goat ... MCM4(4173)

Immunogen

The epitope recognized by PLA0039 maps to a region between residues 325 and 375 of human Minichromosome Maintenance 4 using the numbering given in entry NP_005905.2 (GeneID 4173).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

The MCM (Mini-Chromosome Maintenance) complex is a key component of the pre-replication complex involved in replication licensing which restricts DNA replication to only once per cell cycle. The MCM complex is a heterohexamer of MCM2, MCM3, MCM4, MCM5, MCM6, and MCM7. MCM4 is part of the core MCM complex that includes MCM4, MCM6, and MCM7. The core complex possesses DNA helicase activity. MCM4 is phosphorylated by CDC2 kinase resulting in a reduction of helicase activity and chromatin binding.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spencer W Luebben et al.
Nucleic acids research, 42(9), 5605-5615 (2014-03-05)
Accumulating evidence suggests that dormant DNA replication origins play an important role in the recovery of stalled forks. However, their functional interactions with other fork recovery mechanisms have not been tested. We previously reported intrinsic activation of the Fanconi anemia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service