Skip to Content
Merck
All Photos(1)

Key Documents

P4905

Sigma-Aldrich

DL-Phenylalanine

suitable for cell culture, suitable for insect cell culture

Synonym(s):

(±)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
165.19
Beilstein:
3198807
EC Number:
MDL number:
UNSPSC Code:
12352200

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

mp

266-267 °C (dec.) (lit.)

SMILES string

NC(Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service