P3250
Papain from papaya latex
lyophilized powder, 0.5-2.0 units/mg solid
Synonym(s):
Papainase
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
lyophilized powder
specific activity
0.5-2.0 units/mg solid
mol wt
23.4 kDa
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Papain papaya latex, product P3250, has been used to hydrolyze chitosan to determine whether chitosan can be used in food preservation. It is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.
Used to produce Fab fragments of antibodies. Also used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.
Biochem/physiol Actions
A cysteine protease that cleaves peptide bonds of basic amino acids, leucine, or glycine.
pH optimum 6.0-7.0
Also hydrolyzes esters and amides.
pH optimum 6.0-7.0
Also hydrolyzes esters and amides.
Papain papaya latex has antifungal activity against C. albicans. It is a cysteine protease that cleaves peptide bonds of basic amino acids, leucine, or glycine.
. The pH optimum is 6.0-7.0
. Papain hydrolyzes esters and amides.
. The pH optimum is 6.0-7.0
. Papain hydrolyzes esters and amides.
Unit Definition
One unit will hydrolyze 1.0 micromole of N-alpha-benzoyl-L-arginine ethyl ester (BAEE) per minute at pH 6.2 at 25 deg C
Other Notes
View more information on papain at www.sigma-aldrich.com/enzymeexplorer.
inhibitor
Product No.
Description
Pricing
substrate
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J Rhoades et al.
Applied and environmental microbiology, 66(1), 80-86 (2000-01-05)
The objective of this study was to determine whether chitosan (poly-beta-1,4-glucosamine) and hydrolysates of chitosan can be used as novel preservatives in foods. Chitosan was hydrolyzed by using oxidative-reductive degradation, crude papaya latex, and lysozyme. Mild hydrolysis of chitosan resulted
H Guo et al.
The Journal of physiology, 537(Pt 1), 115-123 (2001-11-17)
1. It has been proposed that the activation of NMDA receptors and upregulation of protein kinase C (PKC) underlie the exaggerated and persistent pain experienced in the inflammatory state. However, there is no direct evidence to show that inflammation alters
Zonghui Dai
Contrast media & molecular imaging, 2022, 2869707-2869707 (2022-06-11)
The aim of the study was to study the protective effect of the Rhizoma Drynariae-Epimedium formula on osteoarthritis in rats and to explore its mechanism. Fifty SD rats were randomly divided into 5 groups, namely, the control group, model group
H Cai et al.
Analytical chemistry, 70(3), 580-589 (1998-02-01)
The third member of a new family of single-isomer charged cyclodextrins, the sodium salt of heptakis(2,3-dimethyl-6-sulfato)-β-cycldextrin, has been synthesized, characterized, and used for the capillary electrophoretic separation of the enantiomers of neutral, acidic, basic, and zwitterionic analytes. Though heptakis(2,3-dimethyl-6-sulfato)-β-cyclodextrin complexes
Yonghua Bi et al.
Journal of vascular and interventional radiology : JVIR, 23(11), 1529-1536 (2012-10-30)
The objective of this study was to determine the possibility of creating a novel animal model of abdominal aortic aneurysm (AAA) in rabbits by the periarterial application of papain. Twelve New Zealand white rabbits were randomized into two groups: (1)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service