Skip to Content
Merck
All Photos(1)

Documents

P2289

Sigma-Aldrich

Endoproteinase Lys-C from Lysobacter enzymogenes

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

Quality Level

analyte chemical class(es)

amino acids

packaging

vial of ≥3.0 units

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Endoproteinase Lys-C is an enzyme that preferentially cleaves at the carboxyl side of lysine residues.

Application

Endoproteinase Lys-C from Lysobacter enzymogenes is useful in the determination of primary structures of proteins. It has been used in a study to investigate the evidence for trisulfide bonds in a recombinant variant of a human IgG2 monoclonal antibody.
Endoproteinase Lys-C has been used for the digestion monoclonal antibodies for disulfide bond assignment.

Biochem/physiol Actions

Endoproteinase Lys-C inactivates the enzyme inositol monophosphatase by cleaving it at a single site directly after Lys 36.

Unit Definition

One unit will hydrolyze 1.0 μmole of N-p-tosyl-Gly-Pro-Lys p-nitroanilide per min at pH 7.7 at 25 °C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiromi Ito et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 806(1), 11-17 (2004-05-20)
Formation of covalently bound protein adducts with 2-arylpropionic acids (2-APAs) has been proposed as a possible explanation for hypersensitivity and toxic responses to chiral carboxylic acid drugs. To identify the cellular proteins chemically modified with optically active (S)-ibuprofen, we generate
The effects of substrates, products and other ligands on the susceptibility ofinositol monophosphatase to proteolysis by endoprotease lys-C.
Gee NS
FEBS Journal, 284, 95-97 (1991)
Wei Zhang et al.
Analytical biochemistry, 311(1), 1-9 (2002-11-21)
Recombinant monoclonal antibodies (mAbs) are an emerging therapeutic area. However, there are few reports on disulfide bond assignment of recombinant mAbs. This work describes the complete disulfide bond assignment of a recombinant immunoglobulin G4 (IgG4) mAb. N-ethylmaleimide (NEM) was used
J G Jacobs et al.
Journal of virology, 85(23), 12537-12546 (2011-09-16)
Classical scrapie is a prion disease in sheep and goats. In sheep, susceptibility to disease is genetically influenced by single amino acid substitutions. Genetic breeding programs aimed at enrichment of arginine-171 (171R) prion protein (PrP), the so-called ARR allele, in
Pavlo Pristatsky et al.
Analytical chemistry, 81(15), 6148-6155 (2009-07-14)
The hinge region of human IgG2 contains four cysteine residues involved in disulfide linkages between the heavy chains, as well as the heavy and light chains. These linkages provide the fundamental framework of three distinct IgG2 disulfide isoforms recently described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service