Skip to Content
Merck
All Photos(1)

Key Documents

P0607

Sigma-Aldrich

Anti-Presenilin-2, N-terminal antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AD3L, Anti-AD4, Anti-PS2, Anti-PSEN2, Anti-Presenilin 2 (Alzheimer 4), Anti-STM2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC (f)
WB
clone:
polyclonal
species reactivity:
human, mouse, rat
citations:
1
technique(s):
immunohistochemistry (frozen sections): 2 μg/mL using 4% paraformaldehyde-fixed mouse brain tissue
western blot: 1-2 μg/mL using mouse or rat brain extracts

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 28 kDa

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry (frozen sections): 2 μg/mL using 4% paraformaldehyde-fixed mouse brain tissue
western blot: 1-2 μg/mL using mouse or rat brain extracts

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PSEN2(5664)
mouse ... Psen2(19165)
rat ... Psen2(81751)

Looking for similar products? Visit Product Comparison Guide

Specificity

Detects an N-terminal proteolytic fragment. Additional bands are observed, probably corresponding to aggregated, modified, or intact PS-2.

Immunogen

synthetic peptide corresponding to amino acids 7-24 of the N-terminal of human presenilin-2 (PS-2). The sequence is the same in mouse and rat.

Physical form

Solution in in 0.05 M sodium phosphate buffer, pH 7.5, containing 0.2% gelatin and 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service