Skip to Content
Merck
All Photos(1)

Key Documents

P0350

Sigma-Aldrich

Procathepsin B Fragment 26-50, rat

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C123H198N34O33S
CAS Number:
Molecular Weight:
2713.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

rat

Quality Level

Assay

≥95% (HPLC)

form

solid

mol wt

2.7 g/mol

composition

Peptide content, ≥70%

UniProt accession no.

storage temp.

−20°C

InChI

1S/C123H198N34O33S/c1-15-67(12)100(156-116(184)87(55-97(168)169)150-119(187)98(65(8)9)154-115(183)86(54-92(129)163)148-113(181)83(51-71-34-38-73(160)39-35-71)144-112(180)82(50-70-27-17-16-18-28-70)145-114(182)85(53-91(128)162)147-106(174)76(32-25-45-133-123(131)132)138-94(165)57-135-103(171)68(13)127)121(189)151-88(60-158)117(185)146-84(52-72-36-40-74(161)41-37-72)111(179)143-80(48-63(4)5)109(177)141-77(30-20-23-43-125)107(175)140-78(31-21-24-44-126)108(176)142-81(49-64(6)7)110(178)152-89(61-191)105(173)137-58-95(166)153-101(69(14)159)122(190)155-99(66(10)11)120(188)149-79(47-62(2)3)104(172)136-56-93(164)134-59-96(167)157-46-26-33-90(157)118(186)139-75(102(130)170)29-19-22-42-124/h16-18,27-28,34-41,62-69,75-90,98-101,158-161,191H,15,19-26,29-33,42-61,124-127H2,1-14H3,(H2,128,162)(H2,129,163)(H2,130,170)(H,134,164)(H,135,171)(H,136,172)(H,137,173)(H,138,165)(H,139,186)(H,140,175)(H,141,177)(H,142,176)(H,143,179)(H,144,180)(H,145,182)(H,146,185)(H,147,174)(H,148,181)(H,149,188)(H,150,187)(H,151,189)(H,152,178)(H,153,166)(H,154,183)(H,155,190)(H,156,184)(H,168,169)(H4,131,132,133)/t67-,68-,69+,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,98-,99-,100-,101-/m0/s1

InChI key

ZFCIHFPJJWSJMS-HGVIYXBOSA-N

Gene Information

rat ... Ctsb(64529)

Amino Acid Sequence

Ala-Gly-Arg-Asn-Phe-Tyr-Asn-Val-Asp-Ile-Ser-Tyr-Leu-Lys-Lys-Leu-Cys-Gly-Thr-Val-Leu-Gly-Gly-Pro-Lys-NH2

Biochem/physiol Actions

Cathepsin B inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Chagas et al.
FEBS letters, 392(3), 233-236 (1996-09-02)
Ten overlapping 15-mer peptides (peptidyl amides) spanning the proregion of rat cathepsin B (residues 1p-60p) were constructed to identify minimal segments having inhibitory activity towards the mature enzyme, that could be used to develop a new generation of peptide-derived inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service