Skip to Content
Merck
All Photos(1)

Key Documents

P0034

Sigma-Aldrich

20S-Protopanaxatriol

Synonym(s):

20(S)-APPT, g-PPT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H52O4
CAS Number:
Molecular Weight:
476.73
MDL number:
UNSPSC Code:
12352200

form

powder

Quality Level

storage temp.

2-8°C

InChI

1S/C30H52O4/c1-18(2)10-9-13-30(8,34)19-11-15-28(6)24(19)20(31)16-22-27(5)14-12-23(33)26(3,4)25(27)21(32)17-29(22,28)7/h10,19-25,31-34H,9,11-17H2,1-8H3/t19-,20+,21-,22+,23-,24-,25-,27+,28-,29+,30-/m0/s1

InChI key

SHCBCKBYTHZQGZ-PHFGEWBZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A ginsenoside metabolite that has been linked to endothelial cell function via the glucocortoid receptor (GR) and the oestrogen receptor (ER).

Application

20S-Protopanaxatriol (g-PPT), a dammarane-type tetracyclic terpene sapogenin, may be used to study its binding to and modulation of cell function via glucocortoid (GR) and oestrogen (ER) receptors.

Biochem/physiol Actions

A metabolites of ginsenoside, protopanaxatriol (g-PPT), could modulate endothelial cell functions through the glucocorticoid receptor (GR) and oestrogen receptor (ER).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaping Ji et al.
Pain, 152(5), 1182-1191 (2011-03-12)
We previously reported that 17β-estradiol (E2) is pronociceptive in a visceral pain model in the rat. Subcutaneously (s.c.) administered E2 reversed the decrease in the colorectal distention (CRD)-evoked visceromotor response produced by ovariectomy (OVx) and CRD-induced nociceptive responses were greater
Jessica Santollo et al.
Physiology & behavior, 97(2), 193-198 (2009-03-04)
Estrogens exert many of their behavioral effects by binding to nuclear estrogen receptor (ER) proteins, ERalpha and ERbeta. Recent studies involving ER knockout mice and selective ER agonists suggest that estradiol's anorexigenic effect is mediated via activation of ERalpha. To
Hoi Hin Kwok et al.
Free radical biology & medicine, 48(3), 437-445 (2009-11-26)
Ginsenosides, the active components of the famous Chinese herb ginseng, have been suggested to possess cardiovascular-protective effects. The mechanism of ginsenosides is believed to be associated with their ability to prevent cellular oxidative stress. The purpose of this study was
So Young Hong et al.
Journal of agricultural and food chemistry, 60(12), 3086-3091 (2012-03-03)
Nitric oxide (NO) produced by endothelial nitric oxide synthase (eNOS) is a fundamental regulator of systemic blood pressure. Ginsenosides from Panax ginseng have been investigated in vitro for the molecular and biochemical mechanisms by which they stimulate NO release in
Rui Liu et al.
Fitoterapia, 81(8), 1079-1087 (2010-07-16)
Effects of protopanaxdiol (PDG) and protopanaxatriol (PTG) types of ginsenosides isolated from the leaves of American ginseng on porcine pancreatic lipase activity were determined in vitro. PDG inhibited the pancreatic lipase activity in a dose-dependent manner at the concentrations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service