Key Documents
OH30058730
Ohaus® Conductivity Meter
Model ST3100C-B, AC/DC input 120 V, US 3-pin plug
About This Item
UNSPSC Code:
41115609
Pricing and availability is not currently available.
AC/DC input
110/120 V, US/Japan plug
material
epoxy shaft
feature
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30058730
conductivity measuring range
0.00-199.9 mS/cm
temperature measuring range
0-100 °C
General description
Among Starter 3100C′s user-friendly features that support ease-of-use are an adjustable standalone electrode holder and large backlit LCD display.
Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results.
Application
Conductivity, total dissolved solids (TDS) measurement
Conductivity, total dissolved solids (TDS), salinity measurement
Features and Benefits
99 measurement memory, automatic and manual endpoint functions, automatic and manual temperature compensation
Automatic temperature compensation, adjustable temperature coefficient
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)