Skip to Content
Merck
All Photos(1)

Documents

OH30057496

Ohaus® pH Meter

Model ST2100-F, AC/DC input 120 V, US 3-pin plug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

material

epoxy shaft

feature

CSA approved

packaging

pack of 1 ea

manufacturer/tradename

Ohaus® 30057496

ORP measuring range

-1999-+1999 mV

pH measuring range

-2-16 pH unit

temperature measuring range

0-100 °C

Looking for similar products? Visit Product Comparison Guide

General description

Easy-to-use and accurate with a simple calibration process, automatic temperature compensation and fast results. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
The built-in electrode holder provides uncomplicated and convenient all-in-one operation while the large LCD display screen clearly displays results.

Application

pH, oxidation-reduction potential (ORP) measurement

Features and Benefits

Auto buffer recognition, two point calibration, quick reference guide
Automatic and manual temperature compensation, auto buffer recognition, calibration icon

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service