Skip to Content
Merck
All Photos(4)

Documents

O9126

Sigma-Aldrich

Oxotremorine sesquifumarate salt

≥98% (HPLC), solid

Synonym(s):

1-(4-[1-Pyrrolidinyl]-2-butynyl)-2-pyrrolidinone, 1-(4-[1-Pyrrolidinyl]2-butynyl)-2-pyrrolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O · 1.5C4H4O4 · xH2O
CAS Number:
Molecular Weight:
380.39 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

mp

102-105 °C (lit.)

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

[H]\C(=C(\[H])C(O)=O)C(O)=O.[H]\C(=C(\[H])C(O)=O)C(O)=O.[H]\C(=C(\[H])C(O)=O)C(O)=O.O=C1CCCN1CC#CCN2CCCC2.O=C3CCCN3CC#CCN4CCCC4

InChI

1S/2C12H18N2O.3C4H4O4/c2*15-12-6-5-11-14(12)10-4-3-9-13-7-1-2-8-13;3*5-3(6)1-2-4(7)8/h2*1-2,5-11H2;3*1-2H,(H,5,6)(H,7,8)/b;;3*2-1+

InChI key

WLYYOFJEBGHKEC-VQYXCCSOSA-N

Gene Information

human ... CHRM2(1129)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oxotremorine sesquifumarate salt is a muscarinic acetylcholine receptor agonist with preference for the M2 receptor.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I B Introini-Collison et al.
Behavioral and neural biology, 57(3), 248-255 (1992-05-01)
Post-training administration of the centrally acting muscarinic agonist oxotremorine (50.0 microgram/kg, ip) facilitated 48-hr retention, in mice, of a one-trial step-through inhibitory avoidance response. Oxotremorine-induced memory facilitation was not prevented by the simultaneous post-training administration of the central beta-adrenoceptor antagonist
P G Gunasekar et al.
Neuroscience research, 49(1), 13-18 (2004-04-22)
Cyanide is generated in neurons and this report examines the two different receptors which mediate cyanide formation in neuronal tissue. An opiate receptor blocked by naloxone increases cyanide production both in rat brain and in rat pheochromocytoma (PC12) cells. A
J D Brioni et al.
Behavioral and neural biology, 50(3), 251-254 (1988-11-01)
The pre-test administration of beta-endorphin (0.05 microgram/mouse), oxotremorine sesquifumarate (5.0 micrograms/mouse), or beta-endorphin plus oxotremorine enhanced retention test performance of a conditioned emotional response in mice. The effects were blocked by scopolamine. Scopolamine had no effect of its own. The
T Reinheimer et al.
American journal of respiratory and critical care medicine, 156(2 Pt 1), 389-395 (1997-08-01)
Human bronchi were incubated in organ baths to measure histamine release. The calcium ionophore A23187 (10 mumol/L; 1 min) stimulated histamine release by 148 +/- 28% (n = 11) above the prestimulation level but was ineffective in epithelium-denuded bronchi. Neither
I Izquierdo et al.
Pharmacology, biochemistry, and behavior, 33(3), 545-548 (1989-07-01)
CD-1 mice were trained in a classically conditioned emotional response paradigm and tested 24 hr later. Exposure to an open field 0 or 1, but not 3 hr after training retroactively interfered with retention of the conditioned emotional response. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service