Skip to Content
Merck
All Photos(1)

Key Documents

N9788

Sigma-Aldrich

NEK6, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

SID6-1512

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€371.00

€371.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€371.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€371.00


Estimated to ship on29 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

114-154 nmol/min·mg

mol wt

~63 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK6(10783)

Biochem/physiol Actions

NEK6 is a serine/threonine kinase that is a member of the Nek family of protein kinases that share an amino-terminal catalytic domain related to NIMA family. NEK6 is a nuclear and cytoplasmic kinase that is required for mitotic progression of human cells. NEK6 is phosphorylated and activated during M phase of the cell cycle.Inhibition of NEK6 function by either overexpression of an inactive NEK6 mutant or elimination of endogenous NEK6 by siRNA arrests cells in M phase and triggers apoptosis.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atish D Choudhury et al.
Cancer research, 77(3), 753-765 (2016-12-03)
In prostate cancer, the development of castration resistance is pivotal in progression to aggressive disease. However, understanding of the pathways involved remains incomplete. In this study, we performed a high-throughput genetic screen to identify kinases that enable tumor formation by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service