Skip to Content
Merck
All Photos(1)

Key Documents

N5513

Sigma-Aldrich

4-Nitrophenyl 2-acetamido-2-deoxy-3-O-β-D-galactopyranosyl-β-D-glucopyranoside

Synonym(s):

p-Nitrophenyl lacto-N-bioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N2O13
CAS Number:
Molecular Weight:
504.44
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(=O)NC1C(OC(CO)C(O)C1OC2OC(CO)C(O)C(O)C2O)Oc3ccc(cc3)N(=O)=O

Substrates

Useful as a chromophoric marker substrate for Lewis blood group-specific fucosyltransferases.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alpha-L-fucosyltransferases in human submaxillary gland and stomach tissues associated with the H, Lea and Leb blood-group characters and ABH secretor status.
M A Chester et al.
Biochemical and biophysical research communications, 34(6), 835-842 (1969-03-31)
Dimitris Koutsioulis et al.
Glycobiology, 18(10), 799-805 (2008-07-19)
In an effort to identify novel endo-alpha-N-acetylgalactosaminidases (endo-alpha-GalNAcases), four potential genes were cloned. Three of the expressed proteins EngEF from Enterococcus faecalis, EngPA from Propionibacterium acnes, and EngCP from Clostridium perfringens were purified and characterized. Their substrate specificity was investigated
Watkins, W.M. et al.
Glycoproteins, 830-830 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service