Skip to Content
Merck
All Photos(1)

Key Documents

N2790

Sigma-Aldrich

Nucleozin

≥98% (HPLC), powder

Synonym(s):

Nucleozin, 1-(2-Chloro-4-nitrophenyl)-4-[(5-methyl-3-phenyl-4-isoxazolyl)carbonyl]-piperazine, [4-(2-Chloro-4-nitro-phenyl)-piperazin-1-yl]-(5-methyl-3-phenyl-isoxazol-4-yl)-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€97.60
25 MG
€370.00

€97.60


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 MG
€97.60
25 MG
€370.00

About This Item

Empirical Formula (Hill Notation):
C21H19ClN4O4
CAS Number:
Molecular Weight:
426.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€97.60


In StockDetails


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >15 mg/mL

storage temp.

2-8°C

SMILES string

Cc1onc(-c2ccccc2)c1C(=O)N3CCN(CC3)c4ccc(cc4Cl)[N+]([O-])=O

InChI

1S/C21H19ClN4O4/c1-14-19(20(23-30-14)15-5-3-2-4-6-15)21(27)25-11-9-24(10-12-25)18-8-7-16(26(28)29)13-17(18)22/h2-8,13H,9-12H2,1H3

InChI key

OWXBJAPOSQSWAO-UHFFFAOYSA-N

Application

Nucleozin may be used in anti-viral (anti-influenza) research to study its pharmacokinetics, metabolism, safety, efficacy and methods of delivery as an influenza A nucleoprotein (a multifunctional, RNA-binding protein necessary for virus replication) targeting drug candidate.[1]

Biochem/physiol Actions

Nucleozin targets influenza A nucleoprotein (NP), a multifunctional, RNA-binding protein necessary for virus replication.
Nucleozin targets influenza A nucleoprotein (NP), a multifunctional, RNA-binding protein necessary for virus replication. Nucleozin is effective in fighting H1N1, H3N2, and H5N1 flu virus strains by inducing the formation of NP aggregates and antagonizing its nuclear accumulation, leading to cessation of viral replication.
Nucleozin targets influenza A nucleoprotein, a multifunctional, RNA-binding protein necessary for virus replication. It induces the formation of nucleoptotein aggregates and inhibits its accumulation, interfering with viral replication. EC50 is in the nM range. Nucleozin is effective in H1N1, H3N2, and H5N1 flu virus strains.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helma Antony et al.
The Analyst, 138(20), 6073-6080 (2013-08-21)
Influenza is a viral pandemic that affects millions of people worldwide. Seasonal variations due to genetic shuffling and antigenic drifts in the influenza viruses have necessitated continual updating of therapeutics. The growing resistance to current influenza drugs has increased demand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service