Skip to Content
Merck
All Photos(1)

Key Documents

N179

Sigma-Aldrich

NS 102

solid

Synonym(s):

6,7,8,9-Tetrahydro-5-nitro-1H-benz[g]indole-2,3-dione 3-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N3O4
CAS Number:
Molecular Weight:
261.23
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

solid

color

yellow

solubility

DMSO: >3 mg/mL
H2O: insoluble
methanol: insoluble

SMILES string

O\N=C1/C(=O)Nc2c3CCCCc3c(cc12)[N+]([O-])=O

InChI

1S/C12H11N3O4/c16-12-11(14-17)8-5-9(15(18)19)6-3-1-2-4-7(6)10(8)13-12/h5,17H,1-4H2,(H,13,14,16)

InChI key

SCDBMLHUXJBJSS-UHFFFAOYSA-N

Biochem/physiol Actions

Competitive glutamate receptor antagonist with high selectivity for the low-affinity [3H] kainate binding site.

Legal Information

Sold under exclusive license from NeuroSearch A/S.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiaki Minami et al.
British journal of pharmacology, 142(4), 679-688 (2004-05-26)
1. Ingestion of a poisonous mushroom Clitocybe acromelalga is known to cause severe tactile pain (allodynia) in the extremities for a month and acromelic acid (ACRO), a kainate analogue isolated from the mushroom, produces selective damage of interneurons of the
Hubert Monnerie et al.
Journal of neuroscience research, 83(6), 944-956 (2006-02-25)
Glutamate is an important regulator of dendrite development that may inhibit, (during ischemic injury), or facilitate (during early development) dendrite growth. Previous studies have reported mainly on the N-methyl-D-aspartate (NMDA) receptor-mediated dendrite growth-promoting effect of glutamate. In this study, we
T H Johansen et al.
European journal of pharmacology, 246(3), 195-204 (1993-08-15)
5-Nitro-6,7,8,9-tetrahydrobenzo[G]indole-2,3-dione-3-oxime (NS-102), a new competitive glutamate receptor antagonist displaced binding to non-N-methyl-D-aspartate (non-NMDA) binding sites with no activity at the NMDA and strychnine-insensitive glycine binding sites. Under experimental conditions in which both high- and low-affinity sites were labelled, NS-102 only
H Kamiya et al.
The Journal of physiology, 509 ( Pt 3), 833-845 (1998-05-23)
1. The effect of a low concentration (1 microM) of kainate (kainic acid; KA) on presynaptic calcium (Ca2+) influx at the Schaffer collateral-commissural (SCC) synapse was examined in rat hippocampal slices. 2. Following selective loading of the presynaptic terminals with
R A Tasker et al.
Canadian journal of physiology and pharmacology, 74(9), 1047-1054 (1996-09-01)
Our objective was to characterize the neurotoxic actions of systemically administered domoic acid on different excitatory amino acid receptors, and to compare the receptor selectivity of domoate with the related compound kainic acid. Groups of mice were injected with various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service