MTCV2250
MTC™ Bio Screw-cap Specimen Vial
polystyrene, capacity 7 mL
Synonym(s):
centrifuge tube clips, laboratory rack, plastic sample tube, sample storage tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear
polystyrene
screw cap
sterility
non-sterile
feature
graduation: no
packaging
pack of 700 ea (with attached screw cap)
manufacturer/tradename
MTC™ Bio V2250
capacity
7 mL
diam. × L
17 mm × 50 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Specimen/Transport Vial, 17x50mm, 7mL, PS, non-graduated, with attached screw-cap, non-sterile, CE, non-toxic, bulk pack of 700
Features and Benefits
- CE marked
- non-cytotoxic
- leak-proof cap
- bulk packaging
Legal Information
MTC is a trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service