MTCP3020
MTC™ Bio Pure.Amp™ PCR 8 Tube Strips
capacity 0.2 mL, seperate dome caps
Synonym(s):
PCR sample storage tubes, sample storage tube, snap closure tube
All Photos(1)
About This Item
Recommended Products
material
(self standing star foot)
clear
polypropylene
sterility
non-sterile
packaging
pack of 120 ea
manufacturer/tradename
MTC™ Bio P3020
capacity
0.2 mL
Related Categories
General description
PCR Tubes Strips of 8 tubes w/ dome caps packaged separately, 120/pk
PureAmp™ 0.2mL PCR strips can be supplied with domed caps or flat caps and can be attached or supplied separately. Optically clear caps and white tubes (for optimal fluorescence reflectivity) are also available for qPCR applications. PureAmp™ 0.1mL PCR strips are intended for qPCR applications and come in Clear, White, and Frosted. Strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Features and Benefits
- RNase, DNase, DNA, and PCR inhibitor free
- 0.2mL or Low Profile 0.1mL options
- Ultra-thin, uniform wall thickness
- White and frosted tubes for enhanced qPCR results
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio