Skip to Content
Merck
All Photos(2)

Key Documents

MAK154

Sigma-Aldrich

Fluorometric Sphingomyelin Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€445.00

€445.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€445.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€445.00


Check Cart for Availability

Request a Bulk Order

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Sphinogmyelin, a sphingolipid consisting of a phospholipid (typically phosphocholine) attached to a ceramide moiety, is a major component of cellular membranes. Sphingomylein is enriched in the exoplasmic leaflet of the cell membrane. Within the cell membrane, sphingomyelin interacts with cholesterol to form lipid rafts. The metabolism of sphingomyelin contributes to cellular signaling via the release of ceramide, a second messenger that participates in multiple cellular signaling pathways. Sphingomyelin accumulates in Niemann-Pick disease due to deficiencies in sphingomyelinase activity.

Suitability

This kit is suitable for the detection of sphingomyelin in a variety of samples such as blood and cell extracts.

Principle

Sphingomyelin is determined by a coupled enzyme assay, in which the hydrolysis of sphingomyelin by sphingomyelinase produces phosphocholine, which results in a fluorescent product (λex =540/λem = 590 nm) proportional to the Sphingomyelin present.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service